Learn More
Sino Biological Recombinant Rhesus NKG2D Protein (aa 78-216, His Tag)
Supplier:  Sino Biological 90164C07B100
Encompass_Preferred
A DNA sequence encoding the rhesus NKG2D (NP_001028061.1) (Phe78-Val216) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-202-1939