Learn More
Sino Biological Recombinant Cynomolgus, Rhesus CD79B Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 90200C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CD79B (XP_005584742.1/NP_001248454.1) (Met1-Asp161) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CD79B sequences are identical.
Catalog No. 50-202-1991