Learn More
Sino Biological Recombinant Rhesus BTLA Protein (hFc Tag)
Supplier:  Sino Biological 90250C02H100
Encompass_Preferred
A DNA sequence encoding the rhesus BTLA (EHH16054.1) (Met1-Leu155) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-202-2075