Learn More
Sino Biological Recombinant Cynomolgus CD25/IL-2R alpha Protein, HPLC-verified
Supplier:  Sino Biological 90265CCCH100
Encompass_Preferred
A DNA sequence encoding the cynomolgus IL2RA (H6WS54) (Met1-Arg213) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-202-2089