Learn More
Sino Biological Cynomolgus EGFR / HER1 / ERBB1 Protein (ECD, His Tag) 50ug
Supplier:  Sino Biological 90285C08H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus EGFR (XP_005549616.1) (Met1-Ser645) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-202-2137