Learn More
Sino Biological Recombinant Cynomolgus, Rhesus Siglec-3/CD33 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90303C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CD33 (EHH59833.1 & EHH30318.1) (Met1-Gly248) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CD33 sequences are identical.
Catalog No. 50-202-2182