Learn More
Sino Biological Recombinant Cynomolgus, Rhesus 4-1BB/CD137 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90847K08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus TNFRSF9 (XP_005544945.1 & NP_001253057.1) (Met1-Gln186) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus TNFRSF9 sequences are identical.
Catalog No. 50-202-2277