Learn More
Chemglass Life Sciences Adapter, Connecting, Inlet, Airfree, Schlenk, 90°
SDP
Supplier:  Chemglass Life Sciences AF007201
Encompass_Preferred
Adapter, Connecting, Inlet, Airfree, Schlenk, 90. Adapter has a #15 o-ring joint with a 0-4mm 1-Arm Chem-Vac Chem-Cap valve. Side arm is 9mm OD medium wall tubing for use with 3/8" Cajon connections. Approx overall height is 100mm. Each adapter is supplied complete with one #116 Viton o-ring.
Catalog No. AF007201