Learn More
Uline Yellow Safety Storage Cabinets 43 x 34 x 65""
Catalog No. NC1790229
Encompass
For aerosols, gasoline and flammable liquids. Unit Weight: 402 lbs. DIMENSIONS: Outside: 43 x 34 x 65" Inside: 39.8 x 30.8 x 61.1" (W x D x H)Shelves (Actual): 39 3/8 x 29" (W x D) Double wall 18-gauge steel. 350 lb. shelf capacity. Adjustable shelves. Sloped shelves safely direct spills away from containers. Recessed paddle handle, can be used with padlock.
Catalog No. NC1790229 Supplier Uline Supplier No. H2219MY
For aerosols, gasoline and flammable liquids. Unit Weight: 402 lbs. DIMENSIONS: Outside: 43 x 34 x 65" Inside: 39.8 x 30.8 x 61.1" (W x D x H)Shelves (Actual): 39 3/8 x 29" (W x D) Double wall 18-gauge steel. 350 lb. shelf capacity. Adjustable shelves. Sloped shelves safely direct spills away from containers. Recessed paddle handle, can be used with padlock.