Learn More
Lab Depot Inc 90mm PETE Membrane Disc, 0.45µm, 30/Pack
SDP
Catalog No. NC3673200
Encompass
Naturally hydrophilic, the PETE membrane disc, 0.45µm, is ideal for blood assays and general filtration. This product also has a broad range of chemical compatibility, including aggressive solvents. It is low protein-binding, ensuring clean results, and has a smooth, thin, glass-like surface.
Catalog No. NC3673200 Supplier Lab Depot Inc Supplier No. 1220702
Naturally hydrophilic, the PETE membrane disc, 0.45µm, is ideal for blood assays and general filtration. This product also has a broad range of chemical compatibility, including aggressive solvents. It is low protein-binding, ensuring clean results, and has a smooth, thin, glass-like surface.