Learn More
Ace Glass, Inc. 90MM POR C FILTR FUNNEL 90Mm Por C Filtr Funnel
SDP
Catalog No. 50979162
Encompass
Funnel with jacket for cooling or heating. All porosities of a given size are priced the same. Hose connection is size C.
Catalog No. 50-979-162 Supplier Ace Glass, Inc. Supplier No. 718526
Funnel with jacket for cooling or heating. All porosities of a given size are priced the same. Hose connection is size C.