Learn More
Ace Glass, Inc. 90mm straight tubes with 25-50 micron fritted discs, for construction of special apparatus.
SDP
Catalog No. 50979204
Encompass_Preferred
90mm straight tubes with 25-50 micron fritted discs, for construction of special apparatus.
Catalog No. 50-979-204 Supplier Ace Glass, Inc. Supplier No. 720576
90mm straight tubes with 25-50 micron fritted discs, for construction of special apparatus.