Learn More
Thermco Products Inc Ultra-Low Freezer bottle thermometer, spirit, -90 deg./20 deg.C, 1.0C div, 145mm, 60ml sand
Catalog No. NC0497398
Encompass_Preferred
Each thermometer is certified, serialized and traceable to NIST.
Catalog No. NC0497398 Supplier Thermco Products Inc Supplier No. ACCULF010S
Each thermometer is certified, serialized and traceable to NIST.