Learn More
Ace Glass, Inc. 90x50mm crystallization dish, cs/18, sp/6, 3140-90
SDP
Catalog No. NC9822320
Encompass
90X50mm Corning crystallization dish made of PYREX glass. Rims are reinforced and fire polished to reduce chipping.
Catalog No. NC9822320 Supplier Ace Glass, Inc. Supplier No. 846512
90X50mm Corning crystallization dish made of PYREX glass. Rims are reinforced and fire polished to reduce chipping.