Learn More
Abcam 9,10-Anthracenediyl-bis(methylene)dimalonic acid, singlet o x ygen assay reagent, 50MG
Catalog No. 502705230
Encompass_Preferred
MW 410.37 g/mol, Purity >90%. Reagent for the assay of singlet oxygen with better characteristics than 9,10-anthracenediyl-bis-dipropionic acid. Photobleached by singlet oxygen to its corresponding endoperoxide.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5230 Supplier Abcam Supplier No. AB14539950MG
MW 410.37 g/mol, Purity >90%. Reagent for the assay of singlet oxygen with better characteristics than 9,10-anthracenediyl-bis-dipropionic acid. Photobleached by singlet oxygen to its corresponding endoperoxide.
The product is subject to the following: Abcam Restricted Use Statement