Learn More
MSC Xuron Diagonal Cutting Plier, 2 mm, 30-12 AWG, 1 ea
Catalog No. 501304627
Encompass
This diagonal cutting plier is designed for precision cutting with an 8.9 mm (0.35 in) jaw length and a 12.2 mm (0.48 in) jaw width. It features an oval head, a rubber handle for comfortable grip, and a cutting capacity suitable for materials up to 2 mm or 30 to 12 AWG.
  • Long handles
  • Ultra precise
  • Lead retainer
  • Return spring
Catalog No. 50-130-4627 Supplier MSC Supplier No. 88356886
This diagonal cutting plier is designed for precision cutting with an 8.9 mm (0.35 in) jaw length and a 12.2 mm (0.48 in) jaw width. It features an oval head, a rubber handle for comfortable grip, and a cutting capacity suitable for materials up to 2 mm or 30 to 12 AWG.
  • Long handles
  • Ultra precise
  • Lead retainer
  • Return spring