Learn More
Cayman Chemical 911 E12nItro Conjugate 100ug
SDP
Catalog No. 502988251
Encompass_Preferred
A nitrated fatty acid; formed from 9(Z),11(E)-CLA upon exposure to acidified nitrite, peroxynitrite, gaseous nitrogen dioxide, or a combination of myeloperoxidase, hydrogen peroxide, and nitrite; formed in LPS-stimulated RAW 264.7 macrophages; has been found in human plasma
Catalog No. 50-298-8251 Supplier Cayman Chemical Supplier No. 30834100 UG
A nitrated fatty acid; formed from 9(Z),11(E)-CLA upon exposure to acidified nitrite, peroxynitrite, gaseous nitrogen dioxide, or a combination of myeloperoxidase, hydrogen peroxide, and nitrite; formed in LPS-stimulated RAW 264.7 macrophages; has been found in human plasma