Learn More
Ace Glass, Inc. 915mm x 14mm OD porosity B sparger tube, 0-5 easy action stopcock top, #7 Ace-Thred side arm
SDP
Catalog No. 50978745
Encompass_Preferred
Catalog No. 50-978-745 Supplier Ace Glass, Inc. Supplier No. 6452145