Learn More
USA Scientific Inc Tough-Tags, 0.94 inch x 0.5 inch, white, on rolls
Catalog No. NC9280333
Encompass
Tough-Tags, small, white labels for 0.5 mL tubes, 0.94 inch L x 0.5 inch W, 1000/roll. For -40C to +121C.
Catalog No. NC9280333 Supplier USA Scientific Inc Supplier No. 91881000
Tough-Tags, small, white labels for 0.5 mL tubes, 0.94 inch L x 0.5 inch W, 1000/roll. For -40C to +121C.