Learn More
eMolecules​ 920-46-7 | Methacryloyl chloride, 1M solution in methylene chloride | Oakwood Chemical | MFCD00000716 | 104.530 | C4H5ClO | 97.000 | CC(=C)C(Cl)=O | 25g | 866714216
Catalog No. 5000163052 Shop All eMolecules Inc. Products
Encompass_Preferred
Methacryloyl chloride, 1M solution in methylene chloride | Oakwood Chemical | 920-46-7 | MFCD00000716 | 104.530 | C4H5ClO | 97.000 | CC(=C)C(Cl)=O | 25g | 866714216
Catalog No. 50-001-63052 Supplier eMolecules​ Supplier No. 866714216
Methacryloyl chloride, 1M solution in methylene chloride | Oakwood Chemical | 920-46-7 | MFCD00000716 | 104.530 | C4H5ClO | 97.000 | CC(=C)C(Cl)=O | 25g | 866714216