Learn More
FUJIFILM Biosciences PBS 1X-Dulbecco's Phosphate Buffered Saline Solution - Liquid
Catalog No. NC0701392
Encompass
PBS 1X Dulbecco's Phosphate Buffered Saline Solution can be used to provide a buffer system to maintain cell culture media in the physiological pH range, or to irrigate, transport, or dilute fluid while maintaining cell tonicity and viability.
Catalog No. NC0701392 Supplier FUJIFILM Biosciences Supplier No. 9240500ML
PBS 1X Dulbecco's Phosphate Buffered Saline Solution can be used to provide a buffer system to maintain cell culture media in the physiological pH range, or to irrigate, transport, or dilute fluid while maintaining cell tonicity and viability.