Learn More
Medchemexpress LLC 93-O17S | 2227008-67-3 | C44H83N3O4S2 | 1 ML
SDP
Catalog No. 5000372836
Encompass_Preferred
93-O17S is an ionizable lipid utilized in the construction of Lipid Nanoparticles (LNP). It is capable of achieving mRNA delivery to the spleen. In vivo studies have demonstrated its ability to deliver luciferase mRNA to the spleen of mice, resulting in detectable mRNA expression in this organ.
  • Ionizable lipid for LNP construction
  • Supports mRNA delivery to the spleen
  • Demonstrates in vivo delivery of luciferase mRNA to mouse spleen
  • High purity (98.0%)
  • Light yellow to yellow oil appearance
Catalog No. 50-003-72836 Supplier Medchemexpress LLC Supplier No. HY15337210MM/1ML
Add to Cart
Add to Cart
93-O17S is an ionizable lipid utilized in the construction of Lipid Nanoparticles (LNP). It is capable of achieving mRNA delivery to the spleen. In vivo studies have demonstrated its ability to deliver luciferase mRNA to the spleen of mice, resulting in detectable mRNA expression in this organ.
  • Ionizable lipid for LNP construction
  • Supports mRNA delivery to the spleen
  • Demonstrates in vivo delivery of luciferase mRNA to mouse spleen
  • High purity (98.0%)
  • Light yellow to yellow oil appearance