Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 93-O17S | 2227008-67-3 | C44H83N3O4S2 | 25 MG
SDP

Catalog No. 5000374552
Encompass_Preferred

93-O17S is an ionizable lipid involved in the construction of Lipid Nanoparticles (LNP) and can achieve mRNA delivery to the spleen. This product is for research use only.

  • Ionizable lipid
  • Involved in the construction of lipid nanoparticles (LNP)
  • Achieves mRNA delivery to the spleen
  • Delivers luciferase mRNA to the spleen of mice
  • Purity: 98.0%
  • Appearance: Oil
  • Color: Light yellow to yellow
  • Soluble in DMSO and ethanol

Catalog No. 50-003-74552 Supplier Medchemexpress LLC Supplier No. 25MGHY153372
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

93-O17S is an ionizable lipid involved in the construction of Lipid Nanoparticles (LNP) and can achieve mRNA delivery to the spleen. This product is for research use only.

  • Ionizable lipid
  • Involved in the construction of lipid nanoparticles (LNP)
  • Achieves mRNA delivery to the spleen
  • Delivers luciferase mRNA to the spleen of mice
  • Purity: 98.0%
  • Appearance: Oil
  • Color: Light yellow to yellow
  • Soluble in DMSO and ethanol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.