Learn More
IW Tremont Co Inc LabExact Filter Binderless glass microfiber 934-AH 1.5um 9.0cm dia. The standard for suspended solids and related measurements (Std Methods 2540D & EPA Method 160.2). Widely used in cell harvesting and RIA scintillation counting. 100/pk
SDP
Supplier:  IW Tremont Co Inc LEAH9000
Encompass_Preferred
LabExact LabExact 934-AH glass microfiber filters are produced from 100% borosilicate glass microfiber without the use of binders or strengthening agents.Fine porosity, fast flow rate with a 1.5 um size particle retention. This material is the standard for suspended solids content and related measurements (Standard Methods 2540D and EPA Method 160.2). Also widely used in cell harvesting applications and RIA scintillation counting. Binderless borosilicate glass microfiber enables use up to 550¿C.Depth filtration categorizes filter media that traps particulate within the random matrix of fibers found within the thickness of the media. The efficiency rating of the filter is characteristic of the complex void space structure that develops as the fibers overlap.Because of the characteristically high flow rates of depth filters, they are generally used in applications such as single and multi process removal of sediment and coarse particulate.
Catalog No. 50-223-7186