Learn More
IW Tremont Co Inc LabExact 934-AH Glass fiber filter, Diameter 4.7cm, prewashed & dried, not tared, TSS or VSS Procedure and related measurements (Std Methods 2540D & EPA Method 160.2). Widely used in cell harvesting and RIA scintillation counting 100/pk
SDP
Supplier:  IW Tremont Co Inc LEAHWD4700
Encompass_Preferred
LabExact Pre-washed and dried Grade 934-AH filters for Total Suspended Solids (TSS) and Volatile Suspended Solids (VSS) are processed using reagent grade water (RGW) under strict controls to wash away extraneous fiber per the requirements of the EPA Standard 2540 C, D & E. Prepared filters are an effective time saver that removes a step from your work flow while maintaining your high levels of quality assurance. Filters are packaged in plastic container, in stacks without weighing pans or bar coding. Available in both 47 and 90mm diameters. Package of 100 washed and ready to use TSS and or VSS Grade 934-AH filters
Catalog No. 50-223-7311