Learn More
Metrohm USA 941 ELUENT PRODUCTION MODULE
SDP
Catalog No. NC2058333 Shop All Metrohm USA Products
Encompass
  • Automated eluent production for ion chromatography
  • Compatible with all Metrohm IC instruments
  • Controlled by MagIC Net software
  • Inline, automated eluent preparation
  • Continuous, maintenance-free operation
  • Operational duration of up to 1 month
  • Integrated 50 mL dosing unit (Dosino)
  • Liquid level monitoring for up to 4 containers
  • Direct connection to ultrapure water system
  • Suitable for processing various chemicals
  • Equipped with 2 USB sockets
  • Equipped with 3 MSB sockets
  • Includes housing with level control, Dosino, and eluent bottle level sensor
  • Provides stable retention times and contamination-free results
  • Offers flexibility in eluent composition and concentration
Catalog No. NC2058333 Supplier Metrohm USA Supplier No. 29410010
Add to Cart
Add to Cart
Request a Quote
  • Automated eluent production for ion chromatography
  • Compatible with all Metrohm IC instruments
  • Controlled by MagIC Net software
  • Inline, automated eluent preparation
  • Continuous, maintenance-free operation
  • Operational duration of up to 1 month
  • Integrated 50 mL dosing unit (Dosino)
  • Liquid level monitoring for up to 4 containers
  • Direct connection to ultrapure water system
  • Suitable for processing various chemicals
  • Equipped with 2 USB sockets
  • Equipped with 3 MSB sockets
  • Includes housing with level control, Dosino, and eluent bottle level sensor
  • Provides stable retention times and contamination-free results
  • Offers flexibility in eluent composition and concentration