Learn More
WELCH MATERIALS INC DoprahTM 9-425 Blue Open Top Ribbed Screw Cap with 9 mm Red PTFE/White Silicone Septa 1.0mm Thick. 100pcs/pk, 10pk/cs
Catalog No. 502093135
Encompass_Preferred
Cap&Septa: 9-425 Blue Open Top Ribbed Screw Cap with 9 mm Red PTFE/White Silicone Septa 1.0mm Thick. 100pcs/pk, 10pk/cs
Catalog No. 50-209-3135 Supplier WELCH MATERIALS INC Supplier No. DR114310
Cap&Septa: 9-425 Blue Open Top Ribbed Screw Cap with 9 mm Red PTFE/White Silicone Septa 1.0mm Thick. 100pcs/pk, 10pk/cs