Learn More
Scientific Commodities Inc 95 DUROMETER PE MICRO MED TUBG
Catalog No. NC9240314
Encompass_Preferred
Micro Tubing, Medical Grade, Polyethylene, I.D. x O.D. x L: 0.045 in. x 0.062 in. x 100 ft.
Catalog No. NC9240314 Supplier Scientific Commodities Inc Supplier No. BB31695PE/7
Micro Tubing, Medical Grade, Polyethylene, I.D. x O.D. x L: 0.045 in. x 0.062 in. x 100 ft.