Learn More
Alkali Scientific 95 kPa Compliant Biohazard Specimen Transport Bags with Absorbent Pad (3"x3"), PP, 0.145 mm Thickness, 12"×15", 500/Case
SDP
Catalog No. 5000663213
Encompass_Preferred
These 95 kPa compliant biohazard specimen transport bags are manufactured from durable polypropylene (PP) and are designed to provide secure, leak resistant containment of biological specimens during transport. Each bag is clearly printed with a biohazard symbol for immediate identification and adherence to biosafety handling requirements. Constructed with a 0.145 mm wall thickness, these bags meet 95 kPa pressure requirements when used as part of a compliant specimen packaging system, making them suitable for transporting liquid and semi liquid specimens. Each bag includes a 3" × 3" absorbent pad to help contain accidental leaks and provide an additional layer of protection during transit. The strong yet flexible PP material supports reliable performance in clinical, laboratory, and diagnostic workflows, ensuring compliant specimen handling and safe transport.
Catalog No. 50-006-63213 Supplier Alkali Scientific Supplier No. SB612X15
These 95 kPa compliant biohazard specimen transport bags are manufactured from durable polypropylene (PP) and are designed to provide secure, leak resistant containment of biological specimens during transport. Each bag is clearly printed with a biohazard symbol for immediate identification and adherence to biosafety handling requirements. Constructed with a 0.145 mm wall thickness, these bags meet 95 kPa pressure requirements when used as part of a compliant specimen packaging system, making them suitable for transporting liquid and semi liquid specimens. Each bag includes a 3" × 3" absorbent pad to help contain accidental leaks and provide an additional layer of protection during transit. The strong yet flexible PP material supports reliable performance in clinical, laboratory, and diagnostic workflows, ensuring compliant specimen handling and safe transport.