Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab 95% Reagent Alcohol
SDP

Catalog No. 22000012
Change view
Click to view available options
Quantity:
30 mL with 15 mL fill
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-000-012 30 mL with 15 mL fill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-012 Supplier StatLab Supplier No. ALREA9530CS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Methanol free denatured alcohol (Methyl alcohol is a known poison that can cause serious health conditions) and suitable for use in clinical laboratory protocols

  • 30 mL vial with 15 mL prefilled solution
  • This denatured alcohol has been formulated using MasterTech 100 percent Reagents Alcohol diluted with 5 percent purified water
  • Ideal for processing and staining protocols that reference 95 percent alcohol in Histology and Cytology laboratories

Specifications

Color Clear
Concentration 95% Alcohol
Product Type Dehydrating Reagent
Quantity 30 mL with 15 mL fill
For Use With (Application) Dehydrant
Reagent Type Alcohol
Form Solution
Format Prefilled Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.