Learn More
Thomas Scientific 95 x 15 mm "Media Saver" Dish
SDP
Catalog No. NC0910974
Encompass
95 mm diameter dish saves 11% media per dish compared to standard dish For use in automated filling equipment Disposable (single use) Polystyrene petri dish for general microbiological use Molded from medical grade virgin polystyrene Optically-clear Radiation sterilized Contents sterile if sleeve is unopened Ventilation ribs allow for free air circulation and reduces condensation during incubation Consistent flatness allows for even media distribution Flared Lid skirt and squared corners for easy one hand operation, especially with gloves Certificates of Sterilization Processing available upon request Traceable by lot and sku number Certificates of Compliance available upon request
Catalog No. NC0910974 Supplier Thomas Scientific Supplier No. 1204U20
95 mm diameter dish saves 11% media per dish compared to standard dish For use in automated filling equipment Disposable (single use) Polystyrene petri dish for general microbiological use Molded from medical grade virgin polystyrene Optically-clear Radiation sterilized Contents sterile if sleeve is unopened Ventilation ribs allow for free air circulation and reduces condensation during incubation Consistent flatness allows for even media distribution Flared Lid skirt and squared corners for easy one hand operation, especially with gloves Certificates of Sterilization Processing available upon request Traceable by lot and sku number Certificates of Compliance available upon request