Learn More
VWR International 95114-012 PerfeCTa® qPCR ToughMix®, Low ROX™ 1250 x 20 μL rxns (10 x 1.25 mL)
Catalog No. NC3825469
Encompass_Preferred
ToughMix additives neutralize PCR inhibitors to ensure reliable assay performance with a spectrum of starting materials including: clinical specimens, plants, soil, environmental or complex food matricesSuperior assay sensitivity and target precision with AccuStart™ II enzyme technology – maximum-yielding, engineered Taq DNA polymerase with stringent, ultrapure antibody hotstart
Catalog No. NC3825469 Supplier VWR International Supplier No. 97065966
ToughMix additives neutralize PCR inhibitors to ensure reliable assay performance with a spectrum of starting materials including: clinical specimens, plants, soil, environmental or complex food matricesSuperior assay sensitivity and target precision with AccuStart™ II enzyme technology – maximum-yielding, engineered Taq DNA polymerase with stringent, ultrapure antibody hotstart