Learn More
Aqua Solutions 9565 500ML/AQUA
SDP
Catalog No. NC9904264
Encompass_Preferred
TOTAL ORGANIC CARBON STANDARD 20.0 PPM PRESERVED WITH HYDROCHLORIC ACID Ph <2
Catalog No. NC9904264 Supplier Aqua Solutions Supplier No. 9565500ML
TOTAL ORGANIC CARBON STANDARD 20.0 PPM PRESERVED WITH HYDROCHLORIC ACID Ph <2