Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 95kPA Biohazard Mailers
GSA_VA

Catalog No. 22130542 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Multiple Container 95kPa Mailer
Multiple Tube 95kPa Mailer
Single Tube 95kPa Mailer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-130-542 Single Tube 95kPa Mailer
22-130-541 Multiple Tube 95kPa Mailer
22-130-540 Multiple Container 95kPa Mailer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
These products do not meet the packaging requirements for Biological Substance Category A materials and do not contain the absorbent materials, cushioning materials, and markings required to ship Biological Substances Category B materials.
Catalog No. 22-130-542

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
These products do not meet the packaging requirements for Biological Substance Category A materials and do not contain the absorbent materials, cushioning materials, and markings required to ship Biological Substances Category B materials.

Includes

Each Dual Mailer comes with a 95kPa pressure tested inner receptacle, an outer fiberboard container and secure, locking Twist & Pull™ end caps. For mailing, shipping and transport compliance, absorbent materials and outer container marking labels must be purchased separately.

For shipments of biological samples by mail and common carriers

Fisherbrand 95kPa Biohazard Dual Mailers are designed for specimen mailing, common carrier shipping and transport within local, regional and national courier networks.

  • For shipments of biological samples by mail and common carriers
  • Accommodates a variety of blood tubes and other routine laboratory containers

Small Single Tube Dual Mailer (Catalog No. 22-130-542)

  • Accommodates a range of standard blood collection tubes from 13 × 75 to 16 × 100mm
  • Inner receptacle dimensions: 0.75 × 5 in. (I.D. × Length)
  • Outer fiberboard mailer dimensions: 1.75 × 7.75 in. (I.D. × Length)
Medium Multiple Tube Dual Mailer (Catalog No. 22-130-541)
  • Two standard blood collection tubes ranging in size from 13 × 75 to 16 × 100mm
  • 15 and 50mL centrifuge tubes, slide specimens and fecal transport vials
  • Inner receptacle dimensions: 2.5 × 4.25 in. (I.D. × Length)
  • Outer Fiberboard mailer dimensions: 2 × 4.75 in. (I.D. × Length)
Large Multiple Container Dual Mailer (Catalog No. 22-130-540)
  • Will accept up to four standard blood collection tubes ranging in size from 13 × 75mm to 16 × 100mm
  • 15 and 50mL centrifuge tubes
  • Slide specimens, fecal transport vials and many other confirmatory test sample containers
  • Inner receptacle dimensions: 2.125 × 4.75 in. (I.D. × Length)
  • Outer fiberboard mailer dimensions: 3.25 × 7 in. (I.D. × Length)

Compliance

Exempt Human Specimen, Exempt Animal Specimen and Biological Substance

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance Exempt Human Specimen, Exempt Animal Specimen and Biological Substance
Applications For transport of biological substances
Type Single Tube 95kPa Mailer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.