Learn More
Biotage 96/BOX GEL FILT PHYTIP COLUMNS
SDP
Catalog No. NC1969886
Encompass
Box of 96 Gel filtration PhyTip columns (1000 µL volume) containing 600 µL of 30K (30K forglobular protein and 10K for dextran) size exclusion resin per column
Catalog No. NC1969886 Supplier Biotage Supplier No. PDR916017
Box of 96 Gel filtration PhyTip columns (1000 µL volume) containing 600 µL of 30K (30K forglobular protein and 10K for dextran) size exclusion resin per column