Learn More
SEQUENCE BIOTECH LLC LTS™Plus 300uL
(compatible with LTS™ Style Pipette Tips including Thermo Scientific™SoftFit-L™) - 5-300uL Working Volume.
Catalog No. NC3311218
Encompass_Preferred
Sequence Biotech introduces the LTS™-Plus 96/384 Channel Benchtop Pipettor.
Designed for use with LTS™ or LTS™ compatible pipette tips. Working volume of 5-300uL.
Key Applications:
ELISA Assays: The Precision Series is ideal for ELISA (Enzyme-Linked Immunosorbent Assays).
Cell Culture/Media Changes: Maintaining healthy cell cultures requires regular media changes, and this pipettor ensures efficient, precise transfers.
PCR Prep and Cleanup: For PCR amplification and subsequent cleanup, this device ensures accurate preparation and removes unwanted components for downstream applications like cloning or sequencing.
Plate-to-Plate Transfers: ensuring accurate and rapid liquid handling across 96-well plates. Ideal for high-throughput labs, it provides precise volume control and seamless operation.
Weighs only 20 lbs.
Catalog No. NC3311218 Supplier SEQUENCE BIOTECH LLC Supplier No. LTSPLUS300
Add to Cart
Add to Cart
Request a Quote
Sequence Biotech introduces the LTS™-Plus 96/384 Channel Benchtop Pipettor.
Designed for use with LTS™ or LTS™ compatible pipette tips. Working volume of 5-300uL.
Key Applications:
ELISA Assays: The Precision Series is ideal for ELISA (Enzyme-Linked Immunosorbent Assays).
Cell Culture/Media Changes: Maintaining healthy cell cultures requires regular media changes, and this pipettor ensures efficient, precise transfers.
PCR Prep and Cleanup: For PCR amplification and subsequent cleanup, this device ensures accurate preparation and removes unwanted components for downstream applications like cloning or sequencing.
Plate-to-Plate Transfers: ensuring accurate and rapid liquid handling across 96-well plates. Ideal for high-throughput labs, it provides precise volume control and seamless operation.
Weighs only 20 lbs.