Learn More
Molecular Devices 96 DEEP SQUARE WELL PLATE RND
Catalog No. NC0188383
Encompass_Preferred
96 Deep Square Well Plate, round bottom, working volume 2000 l; case of 24
Catalog No. NC0188383 Supplier Molecular Devices Supplier No. S1205/DEL
96 Deep Square Well Plate, round bottom, working volume 2000 l; case of 24