Learn More
CELLTREAT Scientific Products 96 Deep Well Storage Plate, 2.0mL, PP, Round Well, Round Bottom, Non-Sterile, 12/CS
SDP
Catalog No. 501535093
Encompass_Preferred
CELLTREAT Scientific Products 96 Deep Well Storage Plate. 96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications. Round Bottom allows use of mixing beads. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Easy to use with automated sample handling instruments. Compatible with multi-channel pipetors. Non-sterile. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged 5/bag, 25/case.
Catalog No. 50-153-5093 Supplier CELLTREAT Scientific Products Supplier No. 229575
Add to Cart
Add to Cart
CELLTREAT Scientific Products 96 Deep Well Storage Plate. 96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications. Round Bottom allows use of mixing beads. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Easy to use with automated sample handling instruments. Compatible with multi-channel pipetors. Non-sterile. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged 5/bag, 25/case.