Learn More
Supply Solutions 96 DEEP WELL 2ML PP ROUND/50PK
Catalog No. NC1456782
Encompass_Preferred
96 Deep well, 2ml/well Polypropylene round well rimless DNA/RNAase free, inner bag of 5. Pack of 50
Catalog No. NC1456782 Supplier Supply Solutions Supplier No. PROVAIR 219020
96 Deep well, 2ml/well Polypropylene round well rimless DNA/RNAase free, inner bag of 5. Pack of 50