Learn More
CELLTREAT Scientific Products 1.1mL 96 Deep Well Storage Plate, PP, V-Bottom, Non-sterile
SDP
Catalog No. 50828742
Encompass_Preferred
CELLTREAT Scientific Products 96 Deep Well Storage Plate. 96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to preventsamples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications. Square to round transition of V-Bottom maximizes working volume. Well volume of 1.1mL, square well shape with a v-bottom. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/bag, 25/case.
Catalog No. 50-828-742 Supplier CELLTREAT Scientific Products Supplier No. 229571
CELLTREAT Scientific Products 96 Deep Well Storage Plate. 96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to preventsamples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications. Square to round transition of V-Bottom maximizes working volume. Well volume of 1.1mL, square well shape with a v-bottom. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/bag, 25/case.