Learn More
Bellco Glass, Inc. 96 deep well plate 1.0mL
SDP
Catalog No. 502330383
Encompass_Preferred
Description96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compoundsPolypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applicationsRound Well with V-Bottom is ideal for vortexing and sample retrieval Pack Description5/BagCase Quantity25SterileYesNon-PyrogenicYesRNase/DNase FreeYesAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneWorking Volume1mLWell ShapeRoundBottom ShapeV-BottomTissue Culture TreatedNoMax Force (RCF)g4000 xg
Catalog No. 50-233-0383 Supplier Bellco Glass, Inc. Supplier No. 229570S00
Description96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compoundsPolypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applicationsRound Well with V-Bottom is ideal for vortexing and sample retrieval Pack Description5/BagCase Quantity25SterileYesNon-PyrogenicYesRNase/DNase FreeYesAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneWorking Volume1mLWell ShapeRoundBottom ShapeV-BottomTissue Culture TreatedNoMax Force (RCF)g4000 xg