Learn More
MICROBIO PRODUCTS LLC 96 Deep Well Plates, 2mL, Round Well, Round Bottom, Clear, 50/case
Catalog No. NC1848001
Encompass_Preferred
UP TO 60% EXTRA YIELD OVER STANDARD PLATES. Features: Culture Growth• High Throughput Protein Expression• Plasmid Purification• Storage• Increased Mixing and Aeration• Increased Culture Production• Decreased Incubation Time• High Throughput, Reproducible Results• Free from Dnase, Rnase, DNA and Detectable PCR Inhibitors Plate Design:• BAFFLED 96 Round Conical Design with Volumes up to 2.0ml• A1 Grade Internal Well Polishing for Minimum Binding• Alphanumeric Frame Coding for Well Identification• Dimensional Conformance to ANSI_SLAS Automated Standards• Raised Wells for Effective Sealing• Precision Molded, Virgin Medical Grade Polypropylene• High Chemical and Temperature ResistantMade of A1-polished ultra-clear medical grade polypropylene. Stable from -8O°C to 120°C with no degradation or sublimation and have been tested to over 6,000 x g providing a safe and strong environment.
Catalog No. NC1848001 Supplier MICROBIO PRODUCTS LLC Supplier No. BPLT2001
Add to Cart
Add to Cart
UP TO 60% EXTRA YIELD OVER STANDARD PLATES. Features: Culture Growth• High Throughput Protein Expression• Plasmid Purification• Storage• Increased Mixing and Aeration• Increased Culture Production• Decreased Incubation Time• High Throughput, Reproducible Results• Free from Dnase, Rnase, DNA and Detectable PCR Inhibitors Plate Design:• BAFFLED 96 Round Conical Design with Volumes up to 2.0ml• A1 Grade Internal Well Polishing for Minimum Binding• Alphanumeric Frame Coding for Well Identification• Dimensional Conformance to ANSI_SLAS Automated Standards• Raised Wells for Effective Sealing• Precision Molded, Virgin Medical Grade Polypropylene• High Chemical and Temperature ResistantMade of A1-polished ultra-clear medical grade polypropylene. Stable from -8O°C to 120°C with no degradation or sublimation and have been tested to over 6,000 x g providing a safe and strong environment.