Learn More
Bellco Glass, Inc. 96 Deep Well Storage Plate
SDP
Catalog No. 502329992
Encompass_Preferred
96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution and is chemically inert for combinatorial chemistry applications.Specifications Square to round transition of VBottom maximizes working volume Pyramid Bottom eases sample retrieval and improves mixing by creating a sloshing effect rather than vortex when agitated Round Bottom allows use of mixing beads Well Volume 0.5mL Well Shape Round Bottom Shape VBottom Case Quantity 25
Catalog No. 50-232-9992 Supplier Bellco Glass, Inc. Supplier No. 229573000
96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution and is chemically inert for combinatorial chemistry applications.Specifications Square to round transition of VBottom maximizes working volume Pyramid Bottom eases sample retrieval and improves mixing by creating a sloshing effect rather than vortex when agitated Round Bottom allows use of mixing beads Well Volume 0.5mL Well Shape Round Bottom Shape VBottom Case Quantity 25