Learn More
Stellar Scientific 96-Well 1.1 mL Deep Well Plate, Square Bottom, 5/PK
SDP
Catalog No. NC1482963
Encompass
HTS-friendly - compatible with most liquid-handling robotsSBS footprint meets ANSI/SLAS 1-2004Flatness tested for reliable automation performanceRaised rims on each well facilitate secure sealing with films or sealing matsOptimum sample mixing and recovery through highly polished wells with radiused cornersUltra-clear for easy sample visualizationCentrifugable to 4000 xgCertified free of DNase, RNase and PCR inhibitors. Sterile plates are also pyrogen freeAlphanumeric indexing molded in on A-H and 1-12 axesAutoclavableStackable
Catalog No. NC1482963 Supplier Stellar Scientific Supplier No. DWP775B00
HTS-friendly - compatible with most liquid-handling robotsSBS footprint meets ANSI/SLAS 1-2004Flatness tested for reliable automation performanceRaised rims on each well facilitate secure sealing with films or sealing matsOptimum sample mixing and recovery through highly polished wells with radiused cornersUltra-clear for easy sample visualizationCentrifugable to 4000 xgCertified free of DNase, RNase and PCR inhibitors. Sterile plates are also pyrogen freeAlphanumeric indexing molded in on A-H and 1-12 axesAutoclavableStackable