Learn More
Thomas Scientific 1.2mL 96-Well Plate, Clear, Square Well, V-Bottom, 100 case
SDP
Catalog No. NC1701284
Encompass_Preferred
Raised rims on each well make for easy and secure sealing with adhesive films, silicone mats, or heat sealing films. Manufactured from virgin polyproplyene and certified to be free of RNase, DNase, and Pyrogen contamination.
Catalog No. NC1701284 Supplier Thomas Scientific Supplier No. 1149J41
Raised rims on each well make for easy and secure sealing with adhesive films, silicone mats, or heat sealing films. Manufactured from virgin polyproplyene and certified to be free of RNase, DNase, and Pyrogen contamination.