Learn More
Waters Corp Waters SPE Cartridge Holder, 96-Flangeless, 1 cc Cartridge
Catalog No. 502037707 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This SPE cartridge holder is designed to batch process 1 cc 96 flangeless SPE cartridges. It specifically holds up to 96 1 cc flangeless SPE cartridges and fits into the 96-position footprint of the manifold. The cartridge and holder assembly is then stacked onto the appropriate collection plate.
  • Designed to hold up to 96 1 cc flangeless SPE cartridges.
  • Fits into the 96-position footprint of the processor manifold.
  • Adapts to the accompanying collection plate for proper installation into the processor.
  • Essential for performing solid-phase extraction at scale.
Catalog No. 50-203-7707 Supplier Waters Corp Supplier No. 186005523
Add to Cart
Add to Cart
This SPE cartridge holder is designed to batch process 1 cc 96 flangeless SPE cartridges. It specifically holds up to 96 1 cc flangeless SPE cartridges and fits into the 96-position footprint of the manifold. The cartridge and holder assembly is then stacked onto the appropriate collection plate.
  • Designed to hold up to 96 1 cc flangeless SPE cartridges.
  • Fits into the 96-position footprint of the processor manifold.
  • Adapts to the accompanying collection plate for proper installation into the processor.
  • Essential for performing solid-phase extraction at scale.