Learn More
Analytical Sales & Services 96 Hole Kapton Plate (0.3mil Liner), 6mm Plate Thickness
Catalog No. 5000806227
Encompass
6mm Thick, Extra-Wide Well Stainless Steel Plate (96 Holes) with 0.3mil Kapton Liner\n\nThis plate is twice as thick as our 96398 plate, and 4x as thick as our standard 1.5mm Kapton plate. For slurry sampling applications requiring even greater volume, the added thickness and extra-wide wells are crucial in containment of the slurry samples to prevent splashout from the well.\n\n• SS plate thickness: 6mm\n• Liner thickness:  0.3mil\n• Well diameter:  8.00mm\n• Well volume: 300 uL/ea\n• SLAS/SBS footprint (1.5 and 3mm plates are slightly smaller)\n• Used to collect Powder X-Ray Diffraction data on polymorph screening samples\n• Designed for both transmission and/or reflection instruments\n\nQty/Pkg: Each
Catalog No. 50-008-06227 Supplier Analytical Sales & Services Supplier No. 966981
6mm Thick, Extra-Wide Well Stainless Steel Plate (96 Holes) with 0.3mil Kapton Liner\n\nThis plate is twice as thick as our 96398 plate, and 4x as thick as our standard 1.5mm Kapton plate. For slurry sampling applications requiring even greater volume, the added thickness and extra-wide wells are crucial in containment of the slurry samples to prevent splashout from the well.\n\n• SS plate thickness: 6mm\n• Liner thickness:  0.3mil\n• Well diameter:  8.00mm\n• Well volume: 300 uL/ea\n• SLAS/SBS footprint (1.5 and 3mm plates are slightly smaller)\n• Used to collect Powder X-Ray Diffraction data on polymorph screening samples\n• Designed for both transmission and/or reflection instruments\n\nQty/Pkg: Each