Learn More
Analytical Sales & Services 96 Hole Kapton Plate (1mil Liner)
Catalog No. 5000804553
Encompass
96 Hole Stainless Steel Plate with Adhesive-Bonded Kapton Film Liner\n• Liner thickness:  1mil\n• SS plate thickness: 1.5mm\n• Well diameter:  6.35mm\n• Used to collect Powder X-Ray Diffraction data on polymorph screening samples\n• Designed for both transmission and/or reflection instruments\n\nQty/Pkg: 10
Catalog No. 50-008-04553 Supplier Analytical Sales & Services Supplier No. 9657710222
96 Hole Stainless Steel Plate with Adhesive-Bonded Kapton Film Liner\n• Liner thickness:  1mil\n• SS plate thickness: 1.5mm\n• Well diameter:  6.35mm\n• Used to collect Powder X-Ray Diffraction data on polymorph screening samples\n• Designed for both transmission and/or reflection instruments\n\nQty/Pkg: 10