Learn More
Microsolv Technology Corporation Inserts, in Racks. Includes 96, Clear, 1000ul Glass Low Volume Precision Inserts with wide conical points. For use in the Micro-Sample Management System. 12/CS.
Catalog No. 502270537
Encompass_Preferred
Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass made for use with the U-2D Micro-Sample Management System. These 1ml Inserts have a wide conical point for fast sample filling to minimize bubble formation and has outer dimensions of 7.72mm x 44.85mm. Preloaded in Racks.
This item is not returnable. View return policy
Catalog No. 50-227-0537 Supplier Microsolv Technology Corporation Supplier No. 952961MLCS
Inserts are 1st Hydrolytic, Class A, TYPE 33 Borosilicate Glass made for use with the U-2D Micro-Sample Management System. These 1ml Inserts have a wide conical point for fast sample filling to minimize bubble formation and has outer dimensions of 7.72mm x 44.85mm. Preloaded in Racks.