Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 96 Channel, 96 Well, Deep Well, or SLAS Tubes or Vials Rack, 19 Gauge, SS Tubes, 42mm Long, 9mm Spacing, Aluminum
Catalog No. NC9869684
Encompass
ASPIRATION MANIFOLD, 96 Channel, 96 Well, Deep Well Plate, or SLAS Tubes or Vials Rack, 19 Gauge, Stainless Steel Tubes, 42mm Long, 9mm Center Spacing, Aluminum, Slide-in Microplate Registration Feature
Catalog No. NC9869684 Supplier V&P Scientific Inc Supplier No. VP 177A1
ASPIRATION MANIFOLD, 96 Channel, 96 Well, Deep Well Plate, or SLAS Tubes or Vials Rack, 19 Gauge, Stainless Steel Tubes, 42mm Long, 9mm Center Spacing, Aluminum, Slide-in Microplate Registration Feature